4-Nitrophenyl phosphate disodium salt (pNNP) 98%

Supplier: Ambeed
Danger

Synonyms: Phosphoric acid 4-nitrophenyl ester disodium salt, 4-Nitrophenyl phosphate disodium salt, PNPP disodium, pNNP

A188124-1G A188124-1MG A188124-5G
103846-666EA 13 USD
103846-666 76721-072 76721-074 77084-394
4-Nitrophenyl phosphate disodium salt (pNNP) 98%
4-Nitrophenyl phosphate disodium salt (pNNP)

Precaución: for Research Use Only

Formula: C₆H₄NNa₂O₆P
Boiling Pt: 457.8 °C (760 mm Hg)
Melting Pt: 300 °C at N/A mmHg
Flash Pt: 230.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00007319
CAS Number: 4264-83-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR