4-Nitrophenyl 4-(pyridin-2-yldisulfanyl)benzyl carbonate 97%

Supplier: Ambeed
Warning

A1144540-100MG A1144540-250MG A1144540-50MG A1144540-5G
103798-408EA 759 USD
103798-408 76673-168 76673-170 76673-172 77783-296
4-Nitrophenyl 4-(pyridin-2-yldisulfanyl)benzyl carbonate 97%
4-Nitrophenyl 4-(pyridin-2-yldisulfanyl)benzyl carbonate

Precaución: for Research Use Only

Formula: C₁₉H₁₄N₂O₅S₂
Storage Temperature: Refrigerator
MDL Number: MFCD32214872
CAS Number: 1151989-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR