2-(4-Nitrophenyl)ethanol 99%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol

A288778-100G A288778-10G A288778-25G A288778-5G
103834-952EA 6 USD
103834-952 76693-188 76693-190 76837-518 77305-544
2-(4-Nitrophenyl)ethanol 99%
2-(4-Nitrophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 177 °C (760 mm Hg)
Melting Pt: 59…62 °C
Flash Pt: 144.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00010202
CAS Number: 100-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR