4-Nitroindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-1H-indole

A190506-1G A190506-25G A190506-5G A190506-100G A190506-10G A190506-250MG
103816-298EA 6 USD
103816-298 103846-900 77084-698 77084-700 77084-702 77236-868 77236-870 77236-872
4-Nitroindole 98%
4-Nitroindole

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 362.6 °C (760 mm Hg)
Melting Pt: 203…207 °C
Flash Pt: 173.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00010056
CAS Number: 4769-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR