4-Nitrobenzyl chloroformate 98%
Supplier: Ambeed
|
Danger
|
A438881-25G
A438881-5G
A438881-10G
A438881-100G
103838-704EA
5
USD
103838-704
76842-454
76842-456
77116-132
77594-368
4-Nitrobenzyl chloroformate 98%
4-Nitrobenzyl chloroformate
Precaución: for Research Use Only
Formula:
C₈H₆ClNO₄ Boiling Pt: 330.8 °C (760 mm Hg) Melting Pt: 32 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007375 CAS Number: 4457-32-3 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...