4-Nitrobenzenesulfonamide 97%

Supplier: Ambeed
Warning

A140319-100G A140319-10G A140319-500G A140319-1G A140319-5G A140319-25G
103802-852EA 9 USD
103802-852 77109-772 77109-774 77109-776 77299-126 77299-128 77315-970
4-Nitrobenzenesulfonamide 97%
4-Nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Boiling Pt: 417.8 °C (760 mm Hg)
Melting Pt: 178…180 °C
Flash Pt: 206.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007937
CAS Number: 6325-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR