4-Nitrobenzene-1,3-diol 95%

Supplier: Ambeed
Warning

A185025-10G A185025-250MG A185025-1G A185025-25G A185025-5G
103815-806EA 28 USD
103815-806 103846-372 76998-818 76998-820 76998-822 77301-938 77301-940 77301-942
4-Nitrobenzene-1,3-diol 95%
4-Nitrobenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₆H₅NO₄
Boiling Pt: 352.8 °C (760 mm Hg)
Flash Pt: 166.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04117945
CAS Number: 3163-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR