4-Nitro-o-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

A111753-100G A111753-10G A111753-25G A111753-500G A111753-5G
103823-718EA 9 USD
103823-718 76821-874 76821-876 76821-878 76821-880 77310-526
4-Nitro-o-phenylenediamine 98%
4-Nitro-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 199…201 °C
Flash Pt: 208.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007724
CAS Number: 99-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR