4-Nitrobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A244916-1000G
A244916-25G
A244916-100G
A244916-1G
A244916-500G
103809-740EA
147
USD
103809-740
103851-446
77090-668
77090-670
77090-672
77113-310
77113-312
77113-314
4-Nitrobenzaldehyde 98%
4-Nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅NO₃ Boiling Pt: 299.6 °C (760 mm Hg) Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007346 CAS Number: 555-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...