4-Nitro-3-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A128234-100MG A128234-250MG A128234-1G A128234-5G
103801-092EA 46 USD
103801-092 76870-618 76870-620 77313-382 77313-384
4-Nitro-3-(trifluoromethyl)benzaldehyde 95%
4-Nitro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
MW: 219.117 g/mol
Boiling Pt: 291.9±40.0 °C (760 mm Hg)
Flash Pt: 130.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185770
CAS Number: 101066-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR