4-Nitro-2-(trifluoromethyl)phenol 97%
Supplier: Ambeed
|
Danger
|
A288443-250MG
A288443-10G
103813-796EA
11
USD
103813-796
103834-938
77002-300
77881-794
4-Nitro-2-(trifluoromethyl)phenol 97%
4-Nitro-2-(trifluoromethyl)phenol
Precaución: for Research Use Only
Formula:
C₇H₄F₃NO₃ Boiling Pt: 259 °C (760 mm Hg) Melting Pt: 105…107 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11100972 CAS Number: 1548-61-4 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...