4-Nitro-1H-pyrazole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Nitro-3-pyrazolecarboxylic acid, 4-Nitropyrazole-3-carboxylic acid
A118088-100G
A118088-10G
A118088-25G
A118088-1G
A118088-5G
103798-990EA
8
USD
103798-990
103824-882
77284-656
77284-658
77284-660
77297-582
77311-672
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₄H₃N₃O₄ MW: 157.085 g/mol Boiling Pt: 509.6 °C (760 mm Hg) Melting Pt: 223…225 °C Flash Pt: 262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090899 CAS Number: 5334-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...