4-Nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A108551-250MG A108551-25G
103797-366EA 10 USD
103797-366 103823-146 77135-830 77684-976
4-Nitro-1H-indazole 95%
4-Nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
Boiling Pt: 383.3 °C (760 mm Hg)
Melting Pt: 199…203 °C
Flash Pt: 185.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00022784
CAS Number: 2942-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR