4-Methylthiophene-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A242451-100MG A242451-1G A242451-250MG A242451-25G
103811-470EA 29 USD
103811-470 103851-282 76976-512 77090-456 77881-637 77881-638
4-Methylthiophene-3-carboxylic acid 97%
4-Methylthiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S
Boiling Pt: 274 °C (760 mm Hg)
Melting Pt: 136…138 °C
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02257717
CAS Number: 78071-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR