4-Methylthiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylthiazole-5-carboxaldehyde, 5-formyl-4-methyl thiazole, 4-Methyl-5-formylthiazole

A210163-1G A210163-5G
103817-982EA 5 USD
103817-982 103848-674 77087-044 77087-046
4-Methylthiazole-5-carbaldehyde 97%
4-Methylthiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.161 g/mol
Melting Pt: 74…78 °C
Flash Pt: 91.8 °C
Storage Temperature: Ambient
MDL Number: MFCD07368277
CAS Number: 82294-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR