4-Methylthiazole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1,3-thiazole-2-carboxylic acid

A148087-250MG A148087-1G
103803-876EA 20 USD
103803-876 103841-990 77286-026 77317-424
4-Methylthiazole-2-carboxylic acid 95%
4-Methylthiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₂S
MW: 143.16 g/mol
Boiling Pt: 317.1 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD07377486
CAS Number: 14542-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR