4-Methylthiazole 98%

Supplier: Ambeed
Danger

A151801-10G A151801-5G A151801-25G
103842-452EA 5 USD
103842-452 76812-218 77299-896 77318-038
4-Methylthiazole 98%
4-Methylthiazole

Precaución: for Research Use Only

Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 133…134 °C (760 mm Hg)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005340
CAS Number: 693-95-8
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR