4-Methyltetrahydro-2H-pyran-4-ol 95%
Supplier: Ambeed
|
Warning
|
A162155-10G
A162155-5G
103805-582EA
34
USD
103805-582
103843-832
76783-912
77319-872
4-Methyltetrahydro-2H-pyran-4-ol 95%
4-Methyltetrahydro-2H-pyran-4-ol
Precaución: for Research Use Only
Formula:
C₆H₁₂O₂ MW: 116.1600 g/mol Boiling Pt: 197.1±8.0 °C (760 mm Hg) Flash Pt: 82.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13193576 CAS Number: 7525-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...