4-Methyltetrahydro-2H-pyran-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A137758-10G A137758-250MG A137758-1G A137758-5G
103802-500EA 33 USD
103802-500 76811-620 76811-622 77298-922 77315-486
4-Methyltetrahydro-2H-pyran-4-carboxylic acid 98%
4-Methyltetrahydro-2H-pyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
Boiling Pt: 256.8 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD11977785
CAS Number: 233276-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR