4-Methyltetrahydro-2H-pyran-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A647260-10G A647260-250MG A647260-1G A647260-5G
103829-222EA 22 USD
103829-222 103840-380 77280-520 77280-522 77308-054 77308-056
4-Methyltetrahydro-2H-pyran-4-carboxylic acid 95%
4-Methyltetrahydro-2H-pyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
Boiling Pt: 256.8 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD11977785
CAS Number: 233276-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR