4-Methyloxan-4-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methyltetrahydro-2H-pyran-4-amine hydrochloride
A215246-10G
A215246-250MG
A215246-1G
A215246-5G
103818-470EA
25
USD
103818-470
103849-174
76975-654
76975-656
77303-400
77303-402
4-Methyloxan-4-amine hydrochloride 97%
4-Methyloxan-4-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO·HCl MW: 151.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13184221 CAS Number: 851389-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...