4-(Methylsulfonyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylsulphonylbenzoic acid

A107704-10G A107704-25G A107704-500G
103823-002EA 5 USD
103823-002 77245-514 77245-516 77245-518
4-(Methylsulfonyl)benzoic acid 97%
4-(Methylsulphonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄S
MW: 200.21 g/mol
Melting Pt: 268…271 °C
Flash Pt: 215.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007564
CAS Number: 4052-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR