4-Methyl-2-quinolinol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Lepidinol, 4-Methylquinolin-2-ol, 2-Hydroxy-4-methylquinoline

A267364-100G A267364-10G A267364-1G A267364-25G A267364-5G
103812-848EA 5 USD
103812-848 103852-746 76772-064 76772-066 77092-478 77304-974 77304-976
4-Methyl-2-quinolinol 97%
4-Methyl-2-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.185 g/mol
Boiling Pt: 346 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00006745
CAS Number: 607-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR