2-Amino-4-methylpyrimidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine
A113264-100G
A113264-10G
A113264-25G
A113264-500G
A113264-5G
103824-044EA
6
USD
103824-044
77009-050
77009-052
77009-054
77009-056
77310-862
2-Amino-4-methylpyrimidine 98%
2-Amino-4-methylpyrimidine
Precaución: for Research Use Only
Formula:
C₅H₇N₃ MW: 109.13 g/mol Melting Pt: 158…160 °C Flash Pt: 149.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006101 CAS Number: 108-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...