4-Methylpyridine-3-sulfonic acid 95%

Supplier: Ambeed
Warning

A333378-1G A333378-5G
103825-636EA 16 USD
103825-636 103836-602 76839-604 76839-606
4-Methylpyridine-3-sulfonic acid 95%
4-Methylpyridine-3-sulfonic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃S
MW: 173.19 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00661349
CAS Number: 4808-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR