4-Methyl-2-pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-methylpyridine, 4-Methylpyridin-2-ol

A292579-1G A292579-5G A292579-10G A292579-25G A292579-100G
103813-944EA 7 USD
103813-944 103835-084 76837-686 76837-688 77289-808 77289-810 77748-571
4-Methyl-2-pyridone 98%
4-Methyl-2-pyridone

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 186…187 °C (760 mm Hg)
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00134298
CAS Number: 13466-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR