4'-Methylpropiophenone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
p-Methylpropiophenone
A913646-1000G
A913646-100G
A913646-25G
A913646-500G
103841-388EA
5
USD
103841-388
77294-222
77294-224
77308-514
77618-546
4'-Methylpropiophenone 97%
4'-Methylpropiophenone
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 238.5 °C (760 mm Hg) Melting Pt: 7.2 °C at N/A mmHg Flash Pt: 97.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009312 CAS Number: 5337-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...