4-Methylpiperidine-4-carbonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A337465-1G A337465-25G A337465-5G A337465-100MG A337465-250MG
103825-748EA 70 USD
103825-748 103836-720 76839-730 76839-732 76839-734 76876-124 76876-126
4-Methylpiperidine-4-carbonitrile hydrochloride 95%
4-Methylpiperidine-4-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂N₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD12406584
CAS Number: 948894-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR