4-Methylpiperidin-4-ol 97%
Supplier: Ambeed
|
Warning
|
A220606-10G
A220606-250MG
A220606-25G
A220606-1G
A220606-5G
103818-860EA
13
USD
103818-860
103849-742
77030-794
77030-796
77030-798
77088-328
77088-330
4-Methylpiperidin-4-ol 97%
4-Methylpiperidin-4-ol
Precaución: for Research Use Only
Formula:
C₆H₁₃NO Boiling Pt: 184.4 °C (760 mm Hg) Flash Pt: 79.471 °C Storage Temperature: Ambient |
MDL Number:
MFCD11100993 CAS Number: 3970-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...