4-Methylpicolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylpicolinic acid, 4-Methylpyridine-2-carboxylic acid

A159299-100G A159299-10G A159299-250MG A159299-1G A159299-25G A159299-5G
103805-242EA 28 USD
103805-242 103843-460 77286-396 77286-398 77286-400 77300-396 77319-334 77319-336
4-Methylpicolinic acid 97%
4-Methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 307.4 °C (760 mm Hg)
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD03731175
CAS Number: 4021-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR