3-Cyano-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile

A205212-25G A205212-5G A205212-100G A205212-10G A205212-500G A205212-1G
103817-618EA 7 USD
103817-618 103848-284 77086-490 77086-492 77274-978 77274-980 77274-982 77302-950
3-Cyano-4-methylpyridine 98%
3-Cyano-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 231.7 °C (760 mm Hg)
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00234272
CAS Number: 5444-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR