4-Methylnicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A158923-100G
A158923-10G
A158923-250MG
A158923-25G
A158923-1G
A158923-5G
103805-192EA
13
USD
103805-192
103843-404
77286-384
77286-386
77286-388
77300-378
77319-264
77319-266
4-Methylnicotinic acid 98%
4-Methylnicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ Melting Pt: 217…221 °C Flash Pt: 137.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128869 CAS Number: 3222-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...