4-Methylisoxazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A177624-1G A177624-100MG A177624-250MG
103807-252EA 198 USD
103807-252 103845-582 77082-968 77096-700 77096-702
4-Methylisoxazole-5-carboxylic acid 97%
4-Methylisoxazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
Boiling Pt: 309.7 °C (760 mm Hg)
Melting Pt: 159…160 °C
Flash Pt: 141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235209
CAS Number: 261350-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR