p-Tolylacetonitrile 98%

Supplier: Ambeed
Warning

A154212-100G A154212-500G A154212-25G
103842-796EA 5 USD
103842-796 77273-072 77273-074 77273-076 77318-494 77318-496
p-Tolylacetonitrile 98%
p-Tolylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₉N
Boiling Pt: 242…243 °C (760 mm Hg)
Melting Pt: 18 °C
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00001922
CAS Number: 2947-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR