p-Toluic acid 98%
Supplier: Ambeed
|
Warning
|
A120071-100G
A120071-10G
A120071-25G
A120071-500G
A120071-1000G
103808-214EA
23
USD
103808-214
76822-354
76822-356
76822-358
76822-360
77297-728
p-Toluic acid 98%
p-Toluic acid
Precaución: for Research Use Only
Formula:
C₈H₈O₂ Boiling Pt: 275.3 °C (760 mm Hg) Melting Pt: 179…182 °C Flash Pt: 124.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002565 CAS Number: 99-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...