4-Methylresorcinol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylbenzene-1,3-diol, 2,4-Dihydroxytoluene

A166560-250MG A166560-1G A166560-25G A166560-5G
103806-076EA 13 USD
103806-076 103844-338 77191-292 77320-598 77320-600 77320-602
4-Methylresorcinol 97%
4-Methylresorcinol

Precaución: Sealed in dry, at room temperature.

Formula: C₇H₈O₂
MW: 124.137 g/mol
Boiling Pt: 264 °C (760 mm Hg)
Melting Pt: 104…108 °C
Flash Pt: 130.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00016452
CAS Number: 496-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR