4-Methyl-5-vinylthiazole 97%

Supplier: Ambeed
Warning

Synonyms: Vinylsulphurol

A276144-25G A276144-5G
103813-296EA 9 USD
103813-296 103853-230 77093-080 77093-082
4-Methyl-5-vinylthiazole 97%
4-Methyl-5-vinylthiazole

Precaución: for Research Use Only

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 185.3 °C (760 mm Hg)
Melting Pt: 15 °C
Flash Pt: 70.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005337
CAS Number: 1759-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR