4-Methyl-5-nitro-1H-indole 97%

Supplier: Ambeed
Warning

A194438-100MG A194438-1G A194438-250MG A194438-5G
103816-658EA 99 USD
103816-658 103847-268 77111-918 77111-920 77111-922 77111-924
4-Methyl-5-nitro-1H-indole 97%
4-Methyl-5-nitro-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
Boiling Pt: 359.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11035915
CAS Number: 165250-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR