2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester

A184040-250MG A184040-1G
103815-704EA 107 USD
103815-704 103846-264 77029-564 77083-876
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester 95%
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈BN₃O₂
MW: 235.09 g/mol
Boiling Pt: 409.7±47.0 °C (760 mm Hg)
Flash Pt: 201.6 °C
Storage Temperature: Freezer
MDL Number: MFCD18072554
CAS Number: 944401-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR