4-Methyl-4H-1,2,4-triazole 97%

Supplier: Ambeed
Warning

A134207-100MG A134207-1G A134207-250MG A134207-5G
103801-984EA 38 USD
103801-984 76870-896 77314-706 77670-922 77670-924
4-Methyl-4H-1,2,4-triazole 97%
4-Methyl-4H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₃H₅N₃
MW: 83.09 g/mol
Boiling Pt: 175 °C (760 mm Hg)
Melting Pt: 90…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00490021
CAS Number: 10570-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR