4-Methyl-4-(trichloromethyl)cyclohexa-2,5-dienone 97%

Supplier: Ambeed
Warning

A101565-10G A101565-25G
103821-716EA 5 USD
103821-716 77257-248 77257-250
4-Methyl-4-(trichloromethyl)cyclohexa-2,5-dienone 97%
4-Methyl-4-(trichloromethyl)cyclohexa-2,5-dienone

Precaución: for Research Use Only

Formula: C₈H₇Cl₃O
MW: 225.5 g/mol
Boiling Pt: 289 °C (760 mm Hg)
Flash Pt: 120.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001590
CAS Number: 3274-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR