4-Methyl-3-phenylpyridine 97%

Supplier: Ambeed
Warning

A268121-100MG A268121-250MG A268121-1G
103812-896EA 114 USD
103812-896 103852-798 77289-388 77289-390 77304-998
4-Methyl-3-phenylpyridine 97%
4-Methyl-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
Boiling Pt: 281.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114238
CAS Number: 19352-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR