4-Methyl-3-oxopentanenitrile 97%

Supplier: Ambeed
Danger

Synonyms: 4-Methyl-3-oxovaleronitrile, Isobutanoylacetonitrile

A456975-1G A456975-5G A456975-10G A456975-250MG
103827-868EA 9 USD
103827-868 103838-944 76842-806 76842-808 77291-756 77291-758
4-Methyl-3-oxopentanenitrile 97%
4-Methyl-3-oxopentanenitrile

Precaución: for Research Use Only

Formula: C₆H₉NO
MW: 111.142 g/mol
Boiling Pt: 160 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05664144
CAS Number: 29509-06-6
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR