4-Methyl-3-oxo-N-phenylpentanamide 98%

Supplier: Ambeed
Warning

A171305-100G A171305-10G A171305-1G A171305-25G A171305-5G
103806-622EA 18 USD
103806-622 103844-896 76998-204 76998-206 77301-116 77301-118 77301-120
4-Methyl-3-oxo-N-phenylpentanamide 98%
4-Methyl-3-oxo-N-phenylpentanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.25 g/mol
Boiling Pt: 384.4 °C (760 mm Hg)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00137795
CAS Number: 124401-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR