3-Nitro-4-picoline 97%

Supplier: Ambeed
Danger

A161180-10G A161180-25G
103805-460EA 13 USD
103805-460 103843-698 77110-704 77300-520
3-Nitro-4-picoline 97%
3-Nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
Boiling Pt: 234.9 °C (760 mm Hg)
Melting Pt: 24…28 °C
Flash Pt: 108.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051829
CAS Number: 5832-44-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR