4-Methyl-3-nitropyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A115913-100G A115913-10G A115913-250MG A115913-25G A115913-1G A115913-5G
103798-648EA 9 USD
103798-648 103824-512 77284-512 77284-514 77284-516 77284-518 77311-334 77311-336
4-Methyl-3-nitropyridin-2(1H)-one 97%
4-Methyl-3-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.12 g/mol
Boiling Pt: 352.5 °C (760 mm Hg)
Melting Pt: 229…232 °C
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00955770
CAS Number: 21901-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR