3-Nitro-p-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitrophenol, 1-Hydroxy-4-methyl-3-nitrobenzene

A211029-100G A211029-10G A211029-500G A211029-25G A211029-5G
103848-760EA 5 USD
103848-760 76770-512 76770-514 76770-516 77087-162 77303-166
3-Nitro-p-cresol 98%
3-Nitro-p-cresol

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 276 °C (760 mm Hg)
Melting Pt: 78…81 °C
Flash Pt: 125.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00007244
CAS Number: 2042-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR