3-Nitro-p-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitrobenzoic acid

A190163-25G A190163-100G A190163-10G A190163-500G A190163-1000G
103809-236EA 98 USD
103809-236 77084-654 77236-858 77236-860 77236-862 77302-214
3-Nitro-p-toluic acid 98%
3-Nitro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 187…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00007174
CAS Number: 96-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR