4-Methyl-5-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitro-1H-pyrazole

A194569-100MG A194569-10G A194569-250MG A194569-1G
103816-668EA 21 USD
103816-668 103847-278 77012-460 77012-462 77012-464 77085-204
4-Methyl-5-nitro-1H-pyrazole 97%
4-Methyl-5-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
Boiling Pt: 317.2±22.0 °C (760 mm Hg)
Flash Pt: 145.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08685901
CAS Number: 38858-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR