4-Methyl-1-tetralone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3,4-dihydronaphthalen-1(2H)-one

A137523-1G A137523-5G
103802-466EA 15 USD
103802-466 77315-436 77315-438
4-Methyl-1-tetralone 97%
4-Methyl-1-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 95 °C (0.75 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001691
CAS Number: 19832-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR